Learn More
Medchemexpress LLC MOG (35-55), human | 163158-19-8 | 98.9% | 2591.99 | 1 MG
SDP
Catalog No. 5000424225
Encompass_Preferred
MOG (35-55), human is a component of CNS myelin. It differs from mMOG (35-55) by a proline for serine substitution at position 42. This peptide is immunogenic via binding to H-2b class II MHC and is recognized by T cells, but it is not encephalitogenic. It shows only partial cross-reactivity with mMOG35-55 and induces minimal clinical signs of EAE compared to the rodent peptide.
  • Component of CNS myelin.
  • Immunogenic via binding to H-2b class II MHC.
  • Recognized by T cells.
  • Induces minimal clinical signs of EAE relative to the rodent peptide.
  • Not encephalitogenic.
  • Partially cross-reactive with mMOG35-55.
Catalog No. 50-004-24225 Supplier Medchemexpress LLC Supplier No. HYP24591MG
MOG (35-55), human is a component of CNS myelin. It differs from mMOG (35-55) by a proline for serine substitution at position 42. This peptide is immunogenic via binding to H-2b class II MHC and is recognized by T cells, but it is not encephalitogenic. It shows only partial cross-reactivity with mMOG35-55 and induces minimal clinical signs of EAE compared to the rodent peptide.
  • Component of CNS myelin.
  • Immunogenic via binding to H-2b class II MHC.
  • Recognized by T cells.
  • Induces minimal clinical signs of EAE relative to the rodent peptide.
  • Not encephalitogenic.
  • Partially cross-reactive with mMOG35-55.