Learn More
AnaSpec MOG(35-55) MOUSE RAT
Supplier:  AnaSpec AS601305
Encompass
MOG (35-55), mouse, rat[MEVGWYRSPFSRVVHLYRNGK]; MW 2582; (5 mg) Myelin oligodendrocyte glycoprotein (MOG) is a member of the immunoglobulin superfamily and is expressed exclusively in the central nervous system (1-3). MOG (35-55) is able to induce autoantibody production and relapsing-remitting neurological disease causing extensive plaque-like demyelination (1-2). Autoantibody response to MOG (35-55) has been observed in multiple sclerosis (MS) patients and MOG (35-55)-induced experimental autoimmune encephalomyelitis (EAE) C57/BL6 mice and Lewis rats (1-4).
Catalog No. 50-844-713