Learn More
Enzo Life Sciences Myelin oligodendrocyte glycoprotein (35-55) (mouse, rat) (1mg). CAS: 163913-87-9
SDP
Supplier:  Enzo Life Sciences ALX162030M001
Encompass_Preferred
Alternative name: MOG (35-55). Formula: C118H177N35O29S. MW: 2582.0. Purity: ≥85% (HPLC). Appearance: White to off-white powder. Solubility: <1mg/ml soluble in water, methanol, DMSO or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-200-7839