Learn More
Carbosynth LLC. MOG (92-106), 5mg, MW: 1823.91, Molecular formula: C80H104N21O27S
SDP
Supplier:  Carbosynth LLC. PMG3968PI5MG
Encompass_Preferred
Synonyms: H-Asp-Glu-Gly-Gly-Tyr-Thr-Cys-Phe-Phe-Arg-Asp-His-Ser-Tyr-Gln-OH, DEGGYTCFFRDHSYQ, Application: Myelin Oligodendrocyte Protein (92-106) Immunogenic Peptide, Storage Temperature: -20°C
Catalog No. 50-168-6668