Learn More
Cayman Chemical MOG PeptIdehumnbovIn 1mg
SDP
Catalog No. 502915168
Encompass_Preferred
An endogenous myelin sheath peptide; reduces cortical and striatal infarct volumes, decreases sensorimotor impairments in tube- and corner turn tests, increases time spent investigating novel odors, and increases the duration of cage mate-directed calls in female, but not male, mice in a MCAO-induced mouse model of ischemic stroke when coupled to HLA-DR2
Catalog No. 50-291-5168 Supplier Cayman Chemical Supplier No. 369061 MG
An endogenous myelin sheath peptide; reduces cortical and striatal infarct volumes, decreases sensorimotor impairments in tube- and corner turn tests, increases time spent investigating novel odors, and increases the duration of cage mate-directed calls in female, but not male, mice in a MCAO-induced mouse model of ischemic stroke when coupled to HLA-DR2