Learn More
Echelon Biosciences Research Labs MOG Protein (35-55), Myelin Oligodendrocyte Glycoprotein peptide fragment (35-55) rat - 1 mg, CAS#: 149635-73-4
SDP
Supplier:  Echelon Biosciences Research Labs 645101MG
Encompass_Preferred
MOG Protein (35-55), Myelin Oligodendrocyte Glycoprotein peptide fragment (35-55) rat - 1mg , Met-Glu-Val-Gly-Trp-Tyr-Arg-Ser-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys-OH Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8771