Learn More
Cayman Chemical MOG35-55mouseratrI 10mg
SDP
Catalog No. 502909625
Encompass_Preferred
An MOG antigen peptide; induces demyelination of the spinal cord, degeneration of the optic nerve, and visual impairments in mice in combination with heat-killed M. tuberculosis and CFA; has been used to induce EAE in animal models of multiple sclerosis in combination with adjuvants
Catalog No. 50-290-9625 Supplier Cayman Chemical Supplier No. 3564410 MG
An MOG antigen peptide; induces demyelination of the spinal cord, degeneration of the optic nerve, and visual impairments in mice in combination with heat-killed M. tuberculosis and CFA; has been used to induce EAE in animal models of multiple sclerosis in combination with adjuvants