Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mogroside II-A | 1613527-65-3 | 10 MG
SDP

Catalog No. 5000417197
Encompass_Preferred

Mogroside II-A is a natural product isolated from the plant Siraitia grosvenorii. It is intended for research use only.

  • Molecular formula: C₄₂H₇₂O₁₄
  • Molecular weight: 801.01
  • Purity: 99.54%
  • Appearance: Solid, white to off-white
  • Storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-17197 Supplier Medchemexpress LLC Supplier No. HYN691510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mogroside II-A is a natural product isolated from the plant Siraitia grosvenorii. It is intended for research use only.

  • Molecular formula: C₄₂H₇₂O₁₄
  • Molecular weight: 801.01
  • Purity: 99.54%
  • Appearance: Solid, white to off-white
  • Storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.