Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mogroside IIA1 | 88901-44-4 | 5 MG
SDP

Catalog No. 5000418722
Encompass_Preferred

Mogroside IIA1 is a triterpenoid glycoside and a nonsugar sweetener. Mogrosides are sweeter than sucrose and exhibit antioxidant, antidiabetic, and anticancer activities.

  • Purity of 99.83%.
  • Appears as a white to off-white solid.
  • Classified as a triterpene from the terpenoid family.
  • Derived from *Siraitia grosvenorii* plants.
  • Intended for research use only.
  • Store at 4°C, protected from light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-18722 Supplier Medchemexpress LLC Supplier No. HYN68555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mogroside IIA1 is a triterpenoid glycoside and a nonsugar sweetener. Mogrosides are sweeter than sucrose and exhibit antioxidant, antidiabetic, and anticancer activities.

  • Purity of 99.83%.
  • Appears as a white to off-white solid.
  • Classified as a triterpene from the terpenoid family.
  • Derived from *Siraitia grosvenorii* plants.
  • Intended for research use only.
  • Store at 4°C, protected from light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.