Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mogroside III | 130567-83-8 | MFCD00238648 | ≥98.0% | 963.15 g/mol | C48H82O19 | 10 MG
SDP

Catalog No. 5000296932
Encompass_Preferred

Mogroside III is a triterpenoid glycoside sold as a purified research reagent and used in studies of carbohydrate metabolism and related pharmacology.

  • Purity ≥98.0%.
  • Molecular weight 963.15 g/mol.
  • Chemical formula C48H82O19.
  • CAS number 130567-83-8.
  • Supplied as a small white to off-white powder in 10 mg packaging.
  • Used in biochemical and pharmacological research, including maltase inhibition studies.

Catalog No. 50-002-96932 Supplier Medchemexpress LLC Supplier No. HYN050010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mogroside III is a triterpenoid glycoside sold as a purified research reagent and used in studies of carbohydrate metabolism and related pharmacology.

  • Purity ≥98.0%.
  • Molecular weight 963.15 g/mol.
  • Chemical formula C48H82O19.
  • CAS number 130567-83-8.
  • Supplied as a small white to off-white powder in 10 mg packaging.
  • Used in biochemical and pharmacological research, including maltase inhibition studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.