Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mogroside III-E | 88901-37-5 | 1 MG
SDP

Catalog No. 5000417946
Encompass_Preferred

Mogroside III-E is a cucurbitane-type compound isolated from Siraitia grosvenorii. It inhibits NO release and exhibits anti-fibrotic activity.

  • Inhibits NO release
  • Exhibits anti-fibrotic activity
  • Purity of 99.30%
  • Molecular weight: 963.15
  • Chemical formula: C48H82O19
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants, Cucurbitaceae, Siraitia grosvenorii

Catalog No. 50-004-17946 Supplier Medchemexpress LLC Supplier No. HYN69281MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mogroside III-E is a cucurbitane-type compound isolated from Siraitia grosvenorii. It inhibits NO release and exhibits anti-fibrotic activity.

  • Inhibits NO release
  • Exhibits anti-fibrotic activity
  • Purity of 99.30%
  • Molecular weight: 963.15
  • Chemical formula: C48H82O19
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants, Cucurbitaceae, Siraitia grosvenorii

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.