Learn More
Medchemexpress LLC Mogroside IIIe | 88901-37-5 | 10 MG
SDP
Catalog No. 5000418117
Encompass_Preferred
Mogroside IIIe is a cucurbitane-type compound isolated from *Siraitia grosvenorii*. It inhibits NO release and possesses anti-fibrotic activity.
  • Cucurbitane-type compound
  • Isolated from *Siraitia grosvenorii*
  • Inhibits NO release
  • Possesses anti-fibrotic activity
  • High purity (99.30%)
  • Soluble in DMSO (100 mg/mL)
  • Available in various in vivo dissolution protocols
Catalog No. 50-004-18117 Supplier Medchemexpress LLC Supplier No. HYN692810MG
Mogroside IIIe is a cucurbitane-type compound isolated from *Siraitia grosvenorii*. It inhibits NO release and possesses anti-fibrotic activity.
  • Cucurbitane-type compound
  • Isolated from *Siraitia grosvenorii*
  • Inhibits NO release
  • Possesses anti-fibrotic activity
  • High purity (99.30%)
  • Soluble in DMSO (100 mg/mL)
  • Available in various in vivo dissolution protocols