Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mogroside IV | 89590-95-4 | 10 MG
SDP

Catalog No. 5000418357
Encompass_Preferred

Mogroside IV is a triterpenoid glycoside isolated from the extracts of Luo Han Guo, functioning as a nonsugar sweetener. Known for being sweeter than sucrose, this compound demonstrates antioxidant, antidiabetic, and anticancer activities. It is supplied as a white to off-white solid with a purity of 98.57%.

  • Triterpenoid glycoside from Siraitia grosvenorii
  • Nonsugar sweetener with higher sweetness than sucrose
  • Exhibits antioxidant, antidiabetic, and anticancer properties
  • Molecular formula: C₅₄H₉₂O₂₄
  • Molecular weight: 1125.29
  • Purity: 98.57%
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-18357 Supplier Medchemexpress LLC Supplier No. HYN694510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mogroside IV is a triterpenoid glycoside isolated from the extracts of Luo Han Guo, functioning as a nonsugar sweetener. Known for being sweeter than sucrose, this compound demonstrates antioxidant, antidiabetic, and anticancer activities. It is supplied as a white to off-white solid with a purity of 98.57%.

  • Triterpenoid glycoside from Siraitia grosvenorii
  • Nonsugar sweetener with higher sweetness than sucrose
  • Exhibits antioxidant, antidiabetic, and anticancer properties
  • Molecular formula: C₅₄H₉₂O₂₄
  • Molecular weight: 1125.29
  • Purity: 98.57%
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month (protect from light)
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.