Learn More
Medchemexpress LLC Mogroside IV | 89590-95-4 | 100 MG
SDP
Catalog No. 5000417826
Encompass_Preferred
Mogroside IV is a triterpenoid glycoside isolated from the extracts of Luo Han Guo, acting as a nonsugar sweetener. Mogrosides are known to be sweeter than sucrose and exhibit antioxidant, antidiabetic, and anticancer activities.
  • Triterpenoid glycoside
  • Nonsugar sweetener
  • Sweeter than sucrose
  • Exhibits antioxidant, antidiabetic, and anticancer activities
  • Isolated from Luo Han Guo extracts
  • Useful in food and metabolic disease research
Catalog No. 50-004-17826 Supplier Medchemexpress LLC Supplier No. HYN6945100MG
Mogroside IV is a triterpenoid glycoside isolated from the extracts of Luo Han Guo, acting as a nonsugar sweetener. Mogrosides are known to be sweeter than sucrose and exhibit antioxidant, antidiabetic, and anticancer activities.
  • Triterpenoid glycoside
  • Nonsugar sweetener
  • Sweeter than sucrose
  • Exhibits antioxidant, antidiabetic, and anticancer activities
  • Isolated from Luo Han Guo extracts
  • Useful in food and metabolic disease research