Learn More
Medchemexpress LLC HY-N6942 5mg Medchemexpress, Mogroside IV-A CAS:88901-41-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN69425MG
Encompass_Preferred
Medchemexpress, HY-N6942 5mg Mogroside IV-A CAS:88901-41-1 Mogroside IV-A, a triterpenoid glycoside isolated from the extracts of Luo Han Guo, is a nonsugar sweetener. Mogrosides are sweeter than sucrose. Mogrosides exhibit antioxidant, antidiabetic and anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2974