Learn More
TARGETMOL CHEMICALS INC MOGROSIDE V 25MG
SDP
Catalog No. 502580448
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mogroside V is a widely used sweetener has in vitro AMPK activating effect it also has anti-inflammatory potential in murine macrophages and a murine ear edema model and has the potential to protect against LPS-induced airway inflammation in a model of ALI. Mogroside V has antioxidant properties and possesses tumor growth inhibitory activity in pancreatic cancer models. purity: 98%
Catalog No. 50-258-0448 Supplier TARGETMOL CHEMICALS INC Supplier No. T297825MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mogroside V is a widely used sweetener has in vitro AMPK activating effect it also has anti-inflammatory potential in murine macrophages and a murine ear edema model and has the potential to protect against LPS-induced airway inflammation in a model of ALI. Mogroside V has antioxidant properties and possesses tumor growth inhibitory activity in pancreatic cancer models. purity: 98%