Learn More
Medchemexpress LLC Mogroside V | 88901-36-4 | MFCD00238649 | 99.8% | 1287.43 | C60H102O29 | 10 MG
SDP
Catalog No. 5000296458
Encompass_Preferred
Mogroside V is a triterpenoid glycoside isolated from Siraitiae Fructus and supplied as a research-grade standard for studies of oxidative stress, mitochondrial function, and metabolic disease models. It reduces intracellular reactive oxygen species and has reported anti-oxidative, anti-diabetic, and anti-carcinogenic effects.
  • High-purity research-grade compound for in vitro and in vivo studies.
  • Characterized by CAS number 88901-36-4 and molecular formula C60H102O29.
  • Available in small-scale quantities suitable for screening and mechanistic assays.
  • Supports studies of oxidative stress, mitochondrial function, and metabolic disease models.
  • Provided with product specification and purity documentation.
Catalog No. 50-002-96458 Supplier Medchemexpress LLC Supplier No. HYN050210MG
Mogroside V is a triterpenoid glycoside isolated from Siraitiae Fructus and supplied as a research-grade standard for studies of oxidative stress, mitochondrial function, and metabolic disease models. It reduces intracellular reactive oxygen species and has reported anti-oxidative, anti-diabetic, and anti-carcinogenic effects.
  • High-purity research-grade compound for in vitro and in vivo studies.
  • Characterized by CAS number 88901-36-4 and molecular formula C60H102O29.
  • Available in small-scale quantities suitable for screening and mechanistic assays.
  • Supports studies of oxidative stress, mitochondrial function, and metabolic disease models.
  • Provided with product specification and purity documentation.