Learn More
Selleck Chemical LLC Mogroside V S3815-25mg
SDP
Catalog No. 503111449
Encompass_Preferred
Mogroside V is a natural cucurbitane glycoside which has a sweetening strength of 250 times that of sucrose and is derived from mature fruit of luo-han-guo (Siraitia grosvenorii monk fruit)
Catalog No. 50-311-1449 Supplier Selleck Chemical LLC Supplier No. S381525MG
Mogroside V is a natural cucurbitane glycoside which has a sweetening strength of 250 times that of sucrose and is derived from mature fruit of luo-han-guo (Siraitia grosvenorii monk fruit)