Learn More
Cayman Chemical Moguisteine, Purity: ≥98%, CAS: 119637-67-1, 100 mg
SDP
Catalog No. 502988838
Encompass_Preferred
A peripherally acting antitussive agent; inhibits the citric acid-induced cough reflex in conscious guinea pigs but not the cough reflex induced by electrical stimulation of the afferent superior laryngeal nerve in anesthetized guinea pigs (ED50s = 5.5 and >20 mg/kg, respectively); prevents BALF eosinophil and neutrophil infiltration in an ovalbumin-sensitized guinea pig model of allergic airway inflammation; inhibits increases in tracheal and bronchial plasma exudation induced by cigarette smoke in anesthetized guinea pigs at 30 mg/kg
Catalog No. 50-298-8838 Supplier Cayman Chemical Supplier No. 30362100 MG
A peripherally acting antitussive agent; inhibits the citric acid-induced cough reflex in conscious guinea pigs but not the cough reflex induced by electrical stimulation of the afferent superior laryngeal nerve in anesthetized guinea pigs (ED50s = 5.5 and >20 mg/kg, respectively); prevents BALF eosinophil and neutrophil infiltration in an ovalbumin-sensitized guinea pig model of allergic airway inflammation; inhibits increases in tracheal and bronchial plasma exudation induced by cigarette smoke in anesthetized guinea pigs at 30 mg/kg