Learn More
Apexbio Technology LLC Moguisteine 119637-67-1 250mg
SDP
Catalog No. 5000088134
Encompass_Preferred
Moguisteine is a non-narcotic compound that modulates the peripheral cough reflex through suppression of sensory nerve endings without direct central nervous system involvement Moguisteine exerts its biological activity primarily through peripheral modulation of sensory nerve signaling pathways Based on these pharmacological properties moguisteine holds research potential in the exploration of peripheral cough pathway regulation and sensory nerve response relevant to respiratory diseases
Catalog No. 50-000-88134 Supplier Apexbio Technology LLC Supplier No. B2115250
Moguisteine is a non-narcotic compound that modulates the peripheral cough reflex through suppression of sensory nerve endings without direct central nervous system involvement Moguisteine exerts its biological activity primarily through peripheral modulation of sensory nerve signaling pathways Based on these pharmacological properties moguisteine holds research potential in the exploration of peripheral cough pathway regulation and sensory nerve response relevant to respiratory diseases