Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Mohs Hardness Test
GREENER_CHOICESCI_ED

Catalog No. S42867 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42867 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.
Catalog No. S42867 Supplier Innovating Science™ Supplier No. IS8703
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.

Students develop vital skills that will help them throughout their studies in earth science. Innovating Science™ Mohs Hardness Test allows students to become familiar with Mohs hardness scale.

Mohs hardness scale was invented by Friedrich Mohs in 1812. Since that moment, the Mohs hardness scale has assisted geologist in determining the identity of minerals.

This kit also provides an opportunity to go out into their local environment and investigate the relative hardness of the minerals around them.

Kit Includes:

  • 1 bag Sample A, 15 Pieces
  • 1 bag Sample B, 15 Pieces
  • 1 bag Sample C, 15 Pieces
  • 1 bag Sample D, 15 Pieces
  • 35 Pieces of Aluminum Foil
  • 35 Scratch Plate
  • 35 Streak Plates

Specifications

Certifications/Compliance Product SDSs
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Science Education
Includes 1 bag Sample A, 15 Pieces, 1 bag Sample B, 15 Pieces, 1 bag Sample C, 15 Pieces, 1 bag Sample D, 15 Pieces, 35 Pieces of Aluminum Foil, 35 Scratch Plate, 35 Streak Plates
Quantity 1
Dimensions (L x W x H) 9 x 6.5 x 2.75 in.
Class Size 15 Lab Groups
Product Type Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.