Learn More
Marketlab, Inc. Moist Mark Laboratory Markers
Description
- Resistant to all solvents, including ethanol and xylene
- Longer lasting
- Fine tip design perfect for small spaces
- Won't dry out like typical lab markers
- Use in urology as a dotting reference marker
Specifications
Specifications
| Type | Permanent Marking Pen |
| Color | Black |
| Quantity | 10 Pk. |