Learn More
Newcomer Supply Moist Mark Plus Pen, 10/pack for painted end of slides and cassettes
SDP
Catalog No. NC9175749
Encompass
Fine-tip (diameter approximately 0.75mm) black ink pen designed for histo and cyto labs. Writes without smearing on both frosted slides along with plastic cassettes. Works well with high pH citrate buffer HIER. Dries quickly.
Catalog No. NC9175749 Supplier Newcomer Supply Supplier No. 6260
Fine-tip (diameter approximately 0.75mm) black ink pen designed for histo and cyto labs. Writes without smearing on both frosted slides along with plastic cassettes. Works well with high pH citrate buffer HIER. Dries quickly.