Learn More
Scientific Industries MOISTURE ANALYZER 110G X .01G
Catalog No. 50728289
Encompass_Preferred
0.1% [110g x 0.01g] 160C Drying general purpose Moisture Analyzer built with computer enhanced strain-gauge, 200W halogen lamp drying oven, External Temperature adjustment, Graphical LCD display, USB, RS232, and PS2 interfaces.
Catalog No. 50-728-289 Supplier Scientific Industries Supplier No. BTS110
0.1% [110g x 0.01g] 160C Drying general purpose Moisture Analyzer built with computer enhanced strain-gauge, 200W halogen lamp drying oven, External Temperature adjustment, Graphical LCD display, USB, RS232, and PS2 interfaces.