Learn More
PCE AMERICAS INC Analytical moisture balance scale with weighing capacity up to 110 g
SDP
Catalog No. NC3654882
Encompass_Preferred
The benchtop scale PCE-MA 100 allows different materials to be weighed and their moisture contents determined. The heating chamber is heated by halogen lamps up to 199 °C. In this chamber, the benchtop scale dries the material to be weighed, and then calculates the moisture content from the weight difference. The benchtop scale can determine the smallest weight differences thanks to a readability of 0.001 g. The results are displayed on the large LCD display. Up to 15 different drying programs can be stored in the memory of the benchtop scale. Accordingly, the benchtop scale is suitable for quality control of wood products, granules, pellets, feed, food, pastes and more. With the included 100 g weight, the benchtop scale can be calibrated at any time.
Catalog No. NC3654882 Supplier PCE AMERICAS INC Supplier No. PCEMA 100
The benchtop scale PCE-MA 100 allows different materials to be weighed and their moisture contents determined. The heating chamber is heated by halogen lamps up to 199 °C. In this chamber, the benchtop scale dries the material to be weighed, and then calculates the moisture content from the weight difference. The benchtop scale can determine the smallest weight differences thanks to a readability of 0.001 g. The results are displayed on the large LCD display. Up to 15 different drying programs can be stored in the memory of the benchtop scale. Accordingly, the benchtop scale is suitable for quality control of wood products, granules, pellets, feed, food, pastes and more. With the included 100 g weight, the benchtop scale can be calibrated at any time.