Learn More
PCE AMERICAS INC Analytical moisture balance scale with a weighing capacity of up to 200 g with a readability of 0.001 g.
SDP
Catalog No. NC3654888
Encompass_Preferred
" Accuracy to 0.01 % The relative humidity meter PCE-MA 200 allows different materials to be weighed and their moisture contents determined. The heating chamber is heated by halogen lamps up to 199 °C. In this chamber, the relative humidity meter dries the material to be weighed, and then calculates the moisture content from the weight difference. The relative humidity meter can determine the smallest weight differences thanks to a readability of 0.001 g. The results are displayed on the large LCD display. Up to 15 different drying programs can be stored in the memory of the relative humidity meter. Accordingly, the relative humidity meter is suitable for quality control of wood products, granules, pellets, feed, food, pastes and more. With the included 100 g weight, the relative humidity meter can be calibrated at any time."
Catalog No. NC3654888 Supplier PCE AMERICAS INC Supplier No. PCEMA 200
" Accuracy to 0.01 % The relative humidity meter PCE-MA 200 allows different materials to be weighed and their moisture contents determined. The heating chamber is heated by halogen lamps up to 199 °C. In this chamber, the relative humidity meter dries the material to be weighed, and then calculates the moisture content from the weight difference. The relative humidity meter can determine the smallest weight differences thanks to a readability of 0.001 g. The results are displayed on the large LCD display. Up to 15 different drying programs can be stored in the memory of the relative humidity meter. Accordingly, the relative humidity meter is suitable for quality control of wood products, granules, pellets, feed, food, pastes and more. With the included 100 g weight, the relative humidity meter can be calibrated at any time."