Learn More
Azenta US Inc Moisture Barrier Seal sheets with 96 adhesive free windows (137 x 76mm); 5 packs of 10 sheets
Catalog No. NC3727711
Encompass
Adhesive sealing film suitable for eukaryotic & bacterial cell culture, long-term incubation, live cell assays, confocal microscopy.Pierceable, peelable, gamma irradiatedGas permeableOptically clear adhesive-free windows
Catalog No. NC3727711 Supplier Azenta US Inc Supplier No. 4TI0516/96
Adhesive sealing film suitable for eukaryotic & bacterial cell culture, long-term incubation, live cell assays, confocal microscopy.Pierceable, peelable, gamma irradiatedGas permeableOptically clear adhesive-free windows