Learn More
Azzota Corp Moisture Determination Balance, Readability: 0.001G, Capacity: 160G, Linearity: +/-0.002G (2Mg), Temp Control: Microprocessor, Heating Time: 1 Min. 50C-100C, Heater Temp: 50C To 160C, Interface: Rs232
Catalog No. 501612880
Encompass_Preferred
Moisture Determination Balance: Readability: 0.001g (1mg), Capacity/Range: 160g, Linearity: +/-0.002g (2mg), Temperature control: Microprocessor, Heating Time: 1 Min. from 50C to 100C, Heater Temperature: 50C to 160C, Interface: RS232, Pan size: Dia. ...
Catalog No. 50-161-2880 Supplier Azzota Corp Supplier No. MTR160
Moisture Determination Balance: Readability: 0.001g (1mg), Capacity/Range: 160g, Linearity: +/-0.002g (2mg), Temperature control: Microprocessor, Heating Time: 1 Min. from 50C to 100C, Heater Temperature: 50C to 160C, Interface: RS232, Pan size: Dia. ...