Learn More
Grainger General Tools Moisture Meter, Digital, Pin Probe, 0 To 50% Range
Catalog No. 502761425
Encompass_Preferred
The General-Purpose Moisture Meter is a microprocessor-based instrument designed to measure the moisture content of various materials including wood fiber, wood products, tobacco, cotton, and paper. It also measures and displays ambient temperature. The unit features 6 in. probes to reach difficult areas and includes a 3.5 ft. cord for versatile operation.
  • Features a wide measuring range of 0% to 50% moisture and high resolution.
  • Equipped with a large 4-digit LCD for clear, easy-to-read data.
  • Includes a data hold function and remote probe for added convenience.
  • Operates on 4 AA batteries (included) and comes with a carrying case.
  • Measures ambient temperature from 14°F to 150°F.
  • Designed to be compact, lightweight, and easy to use.
Catalog No. 50-276-1425 Supplier Grainger Supplier No. 15X981
The General-Purpose Moisture Meter is a microprocessor-based instrument designed to measure the moisture content of various materials including wood fiber, wood products, tobacco, cotton, and paper. It also measures and displays ambient temperature. The unit features 6 in. probes to reach difficult areas and includes a 3.5 ft. cord for versatile operation.
  • Features a wide measuring range of 0% to 50% moisture and high resolution.
  • Equipped with a large 4-digit LCD for clear, easy-to-read data.
  • Includes a data hold function and remote probe for added convenience.
  • Operates on 4 AA batteries (included) and comes with a carrying case.
  • Measures ambient temperature from 14°F to 150°F.
  • Designed to be compact, lightweight, and easy to use.