The Dual Moisture Meter is a versatile digital instrument designed for professional moisture detection in construction and building projects. This dual-function device utilizes both pin and pinless sensing technology to provide accurate measurements in wood and various building materials. It features an integrated LCD display with low battery indicator, data hold capabilities, and an audible alert system that scales in intensity as moisture levels rise to ensure efficient operation.
- Compatible with wood, drywall, concrete, plaster, mortar, cardboard, and stone
- Built-in measurement verification and battery check functions
- Includes protective cap, meter, and replacement pins
- Measures wood moisture from 5% to 50% in pin mode and 0% to 99.9% in pinless mode
- Maximum detection depths of 0.3 in. (pin) and 1 in. (pinless)
- Auto shut-off after 2 min.