Learn More
PCE AMERICAS INC Analytical moisture balance scale with weighing capacity from 0.01 g to 110 g
SDP
Catalog No. NC3654884
Encompass_Preferred
"PCE-MA 110 is an analytical balance or halogen moisture analyzer that functions on the gravimetric or loss on drying (LOD) principle. The moisture analyzer uses the halogen method to dry the material sample. Moisture content is calculated based on the weight difference before and after the drying procedure. Calibration of the moisture analyzer can be performed at any time using the automatic calibration function and the included 100 g weight.Weighing range: 0.01 g … 110 g / 0.0035 … 3.88 ozWeighing resolution or readability: 0.1 g / 10 mgMoisture content measuring range: 0 … 100%Moisture content resolution or readability: 0.1%Adjustable drying time and temperatureMemory stores up to 16 drying profiles"
Catalog No. NC3654884 Supplier PCE AMERICAS INC Supplier No. PCEMA 110
"PCE-MA 110 is an analytical balance or halogen moisture analyzer that functions on the gravimetric or loss on drying (LOD) principle. The moisture analyzer uses the halogen method to dry the material sample. Moisture content is calculated based on the weight difference before and after the drying procedure. Calibration of the moisture analyzer can be performed at any time using the automatic calibration function and the included 100 g weight.Weighing range: 0.01 g … 110 g / 0.0035 … 3.88 ozWeighing resolution or readability: 0.1 g / 10 mgMoisture content measuring range: 0 … 100%Moisture content resolution or readability: 0.1%Adjustable drying time and temperatureMemory stores up to 16 drying profiles"