Learn More
Chemglass Life Sciences Moisture Test Adapter, 45/50 Joint, 250mm
SDP
Catalog No. 50872914
Encompass_Preferred
Moisture Test Adapter, 45/50 Joint, 250mm. Adapter used to construct a simple moisture trap. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint. Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50L.
Catalog No. 50-872-914 Supplier Chemglass Life Sciences Supplier No. CG106309
Moisture Test Adapter, 45/50 Joint, 250mm. Adapter used to construct a simple moisture trap. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint. Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50L.