Learn More
Chemglass Life Sciences Distilling Receiver, 10mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm Glass Stopcock, Approx. 275mm Height x 95mm Width
SDP
Catalog No. CG126305
Encompass_Preferred
Distilling Receiver, 10mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm Glass Stopcock, Approx. 275mm Height x 95mm Width. Similar to CG-1261 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Catalog No. CG126305 Supplier Chemglass Life Sciences Supplier No. CG126305
Distilling Receiver, 10mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm Glass Stopcock, Approx. 275mm Height x 95mm Width. Similar to CG-1261 but with the addition of an overflow tube between the uptake tube and the graduated receiver.