Learn More
Chemglass Life Sciences Joint Size: 24/40, Capacity mL: 10, Stopcock Size: 2mm, Approx. Height x Width (mm): 275 x 125
SDP
Catalog No. CG1261A10
Encompass_Preferred
Graduated distilling receiver with a standard taper outer joint on top and a standard taper lower drip joint. Distance between the receiver and the uptake arm: 10 and 20mL are 100mm center to center; the 50, 100 and 250mL are 150mm center to center. Joint Size: 24/40, Capacity mL: 10, Stopcock Size: 2mm, Approx. Height x Width (mm): 275 x 125.
Catalog No. CG1261A10 Supplier Chemglass Life Sciences Supplier No. CG1261A10
Graduated distilling receiver with a standard taper outer joint on top and a standard taper lower drip joint. Distance between the receiver and the uptake arm: 10 and 20mL are 100mm center to center; the 50, 100 and 250mL are 150mm center to center. Joint Size: 24/40, Capacity mL: 10, Stopcock Size: 2mm, Approx. Height x Width (mm): 275 x 125.