Learn More
Chemglass Life Sciences Joint Size: 45/50, Capacity mL: 250, Stopcock Size: 4mm, Approx. Height x Width (mm): 330 x 196
SDP
Catalog No. CG1261A49
Encompass_Preferred
Graduated distilling receiver with a standard taper outer joint on top and a standard taper lower drip joint. Distance between the receiver and the uptake arm: 10 and 20mL are 100mm center to center; the 50, 100 and 250mL are 150mm center to center. Joint Size: 45/50, Capacity mL: 250, Stopcock Size: 4mm, Approx. Height x Width (mm): 330 x 196.
Catalog No. CG1261A49 Supplier Chemglass Life Sciences Supplier No. CG1261A49
Graduated distilling receiver with a standard taper outer joint on top and a standard taper lower drip joint. Distance between the receiver and the uptake arm: 10 and 20mL are 100mm center to center; the 50, 100 and 250mL are 150mm center to center. Joint Size: 45/50, Capacity mL: 250, Stopcock Size: 4mm, Approx. Height x Width (mm): 330 x 196.