Learn More
Chemglass Life Sciences Distilling Receiver, 10mL, Moisture Test, Dean-Stark, 24/40 Joint Size, Approx. 250mm Height x 90mm Width
SDP
Catalog No. CG125903
Encompass_Preferred
Distilling Receiver, 10mL, Moisture Test, Dean-Stark, 24/40 Joint Size, Approx. 250mm Height x 90mm Width. Similar to CG-1258 but with the addition of an overflow tube between the uptake tube and the graduated receiver.
Catalog No. CG125903 Supplier Chemglass Life Sciences Supplier No. CG125903
Distilling Receiver, 10mL, Moisture Test, Dean-Stark, 24/40 Joint Size, Approx. 250mm Height x 90mm Width. Similar to CG-1258 but with the addition of an overflow tube between the uptake tube and the graduated receiver.