Learn More
Chemglass Life Sciences Moisture Test Adapter, 24/40 Joint, 200mm
SDP
Catalog No. 501531000
Encompass_Preferred
Moisture Test Adapter, 24/40 Joint, 200mm. Adapter used to construct a simple moisture trap. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint. Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50L.
Catalog No. 50-153-1000 Supplier Chemglass Life Sciences Supplier No. CG106305
Moisture Test Adapter, 24/40 Joint, 200mm. Adapter used to construct a simple moisture trap. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint. Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50L.