Learn More
Chemglass Life Sciences Moisture Test Adapter, 14/20 Joint, 100mm
SDP
Catalog No. 501530999
Encompass_Preferred
Moisture Test Adapter, 14/20 Joint, 100mm. Adapter used to construct a simple moisture trap. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint. Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50L.
Catalog No. 50-153-0999 Supplier Chemglass Life Sciences Supplier No. CG106301
Moisture Test Adapter, 14/20 Joint, 100mm. Adapter used to construct a simple moisture trap. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint. Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50L.