Learn More
Cole-Parmer Chromatography Research Supplies Large-capacity moisture traps; 400 mL volume; 1/8" fitting; no base plate
Catalog No. NC1394890 Shop All Cole-Parmer Products
Encompass_Preferred
Traps remove moisture, oils, and dust from carrier gases. Large 400-mL capacity means fewer refillings are required. Adsorbent materials are indicating desiccant and 5-Å molecular sieve. Desiccant changes color from blue to pink when depleted. Easily refill the trap with refill kit 34651-39. The silicone O-ring seals maintain pressures to 100 psi. Traps feature transparent acrylic housing. Measure 17"L x 2" dia. Choose a trap with base plate for benchtop mounting. Shpg wt 3 lb (1.4 kg).
Catalog No. NC1394890 Supplier Cole-Parmer Supplier No. 3465120
Traps remove moisture, oils, and dust from carrier gases. Large 400-mL capacity means fewer refillings are required. Adsorbent materials are indicating desiccant and 5-Å molecular sieve. Desiccant changes color from blue to pink when depleted. Easily refill the trap with refill kit 34651-39. The silicone O-ring seals maintain pressures to 100 psi. Traps feature transparent acrylic housing. Measure 17"L x 2" dia. Choose a trap with base plate for benchtop mounting. Shpg wt 3 lb (1.4 kg).