Learn More
Biolegend MojoSort™ Human anti-APC Nanobeads
Supplier:  Biolegend 480090
Encompass_Preferred
MojoSort™ Human anti-APC Nanobeads Reactivity: Human; Apps: Cell Sep - Pos, Cell Sep - Neg; Size: 1 mL
Catalog No. 50-208-5443