Learn More
Biolegend MojoSort™ Human CD4 Nanobeads
Catalog No. 502080042
Encompass_Preferred
MojoSort™ Human CD4 Nanobeads Reactivity: Human; Apps: Cell Sep - Pos; Size: 10 tests
Catalog No. 50-208-0042 Supplier Biolegend Supplier No. 480013
MojoSort™ Human CD4 Nanobeads Reactivity: Human; Apps: Cell Sep - Pos; Size: 10 tests