Learn More
Biolegend MojoSort™ Human CD4 Naïve T Cell Isolation Kit
Supplier:  Biolegend 480042
Encompass_Preferred
MojoSort™ Human CD4 Naïve T Cell Isolation Kit Reactivity: Human; Apps: Cell Sep - Neg; Size: 200 tests
Catalog No. 50-208-0060