Learn More
Biolegend MojoSort™ Mouse CD4 T Cell Isolation Kit
Catalog No. 502080054
Encompass_Preferred
MojoSort™ Mouse CD4 T Cell Isolation Kit Reactivity: Mouse; Apps: Cell Sep - Neg; Size: 200 tests
Catalog No. 50-208-0054 Supplier Biolegend Supplier No. 480033
MojoSort™ Mouse CD4 T Cell Isolation Kit Reactivity: Mouse; Apps: Cell Sep - Neg; Size: 200 tests