Learn More
Biolegend MojoSort™ Mouse CD45 Nanobeads
Catalog No. 502080050
Encompass_Preferred
MojoSort™ Mouse CD45 Nanobeads Reactivity: Mouse; Apps: Cell Sep - Pos; Size: 20 tests
Catalog No. 50-208-0050 Supplier Biolegend Supplier No. 480027
MojoSort™ Mouse CD45 Nanobeads Reactivity: Mouse; Apps: Cell Sep - Pos; Size: 20 tests