Learn More
Biolegend MojoSort on Columns - Human CD4 Na ve T Cell Isolation Kit Reactivity Human Apps Cell Sep - Neg Size 100 tests
Catalog No. 5000043815
Encompass_Preferred
MojoSort on Columns - Human CD4 Na ve T Cell Isolation Kit Reactivity Human Apps Cell Sep - Neg Size 100 tests
Catalog No. 50-000-43815 Supplier Biolegend Supplier No. 480181
MojoSort on Columns - Human CD4 Na ve T Cell Isolation Kit Reactivity Human Apps Cell Sep - Neg Size 100 tests