Learn More
Biolegend MojoSortTM Mouse Pan B Cell Is
Catalog No. 502721898
Encompass_Preferred
MojoSort™ Mouse Pan B Cell Isolation Kit II; Reactivity: Mouse; Apps: Cell Sep - Neg; Size: 20 tests
Catalog No. 50-272-1898 Supplier Biolegend Supplier No. 480087
MojoSort™ Mouse Pan B Cell Isolation Kit II; Reactivity: Mouse; Apps: Cell Sep - Neg; Size: 20 tests