Learn More
Eurofins Discoverx MOK, active, 10 µg. (14-960)
Supplier:  Eurofins Discoverx 14960
Encompass
N-terminal GST-tagged and C-terminal 6xHis tagged recombinant, human MOK amino acids 1-343. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9488