Learn More
Alkali Scientific CellPro™ DEPC Treated Ultra Pure Water, 500ml
SDP
Catalog No. 502389376
Encompass_Preferred
DEPC Treated Water (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases. Water is usually treated with 0.1% v/v DEPC for at least 2 hours at 37 °C and then autoclaved (at least 15 min) to inactivate traces of DEPC. Inactivation of DEPC in this manner yields CO 2 and ethanol.
Catalog No. 50-238-9376 Supplier Alkali Scientific Supplier No. DC42111
DEPC Treated Water (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases. Water is usually treated with 0.1% v/v DEPC for at least 2 hours at 37 °C and then autoclaved (at least 15 min) to inactivate traces of DEPC. Inactivation of DEPC in this manner yields CO 2 and ethanol.