Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Molasses
SCI_ED

Catalog No. S25753 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
300 mL
Packaging:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25753 300 mL Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25753

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 139-05-9
Molecular Formula C6H12NNaO3S
Molecular Weight (g/mol) 201.22
MDL Number MFCD00003827
InChI Key UDIPTWFVPPPURJ-UHFFFAOYSA-M
Synonym sodium cyclamate, sodium cyclohexylsulfamate, sodium n-cyclohexylsulfamate, cyclamate sodium, cyclamic acid sodium salt, assugrin, ibiosuc, suessette, suestamin, sugarin
PubChem CID 23665706
ChEBI CHEBI:82431
IUPAC Name sodium N-cyclohexylsulfamate
SMILES [Na+].[O-]S(=O)(=O)NC1CCCCC1

Specifications

Density 1.4g/mL
Packaging Solution
Quantity 300 mL
Chemical Name or Material Molasses
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.