Learn More
R&D Systems™ MolBoolean Mouse/Rabbit Assay Kit

Specifications
Specifications
| Species | Mouse |
| Product Line | Novus |
| Content And Storage | Store at -20°C. |
| For Use With (Application) | Primary antibodies are not supplied as part of the MolBoolean kit. The user should include their own antibodies, one of rabbit and one of mouse origin, which have been validated in relevant applications. |
| Quantity | 4.8 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.